siRNA Details
Gene Symbol: Tpm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024345.2 | NP_001019516.1 | 3, 4 | 557 | ||
NM_001301235.1 | NP_001288164.1 | 3, 4 | 557 | |||
XM_008763680.1 | XP_008761902.1 | 3, 4 | 897 | |||
XM_008763681.1 | XP_008761903.1 | 3, 4 | 897 | |||
XM_008763682.1 | XP_008761904.1 | 2, 3 | 275 | |||
GenBank mRNA | BC090009.1 | - | 3, 4 | 478 | 7324151 | |
CF110059.1 | - | 3, 4 | 443 | |||
FQ214609.1 | - | 3, 4 | 492 | |||
FQ214635.1 | - | 3, 4 | 494 | |||
FQ214711.1 | - | 3, 4 | 494 | |||
FQ214816.1 | - | 3, 4 | 473 | |||
FQ214823.1 | - | 3, 4 | 494 | |||
FQ214899.1 | - | 3, 4 | 494 | |||
FQ215016.1 | - | 3, 4 | 457 | |||
FQ215066.1 | - | 3, 4 | 494 | |||
FQ215076.1 | - | 3, 4 | 454 | |||
FQ215143.1 | - | 3, 4 | 494 | |||
FQ215150.1 | - | 3, 4 | 494 | |||
FQ215210.1 | - | 3, 4 | 494 | |||
FQ215302.1 | - | 3, 4 | 494 | |||
FQ215335.1 | - | 3, 4 | 454 | |||
FQ215502.1 | - | 3, 4 | 494 | |||
FQ215601.1 | - | 3, 4 | 494 | |||
FQ215676.1 | - | 3, 4 | 494 | |||
FQ215883.1 | - | 3, 4 | 492 | |||
FQ215920.1 | - | 3, 4 | 492 | |||
FQ216112.1 | - | 3, 4 | 493 | |||
FQ216187.1 | - | 3, 4 | 494 | |||
FQ216228.1 | - | 3, 4 | 494 | |||
FQ216294.1 | - | 3, 4 | 493 | |||
FQ216477.1 | - | 3, 4 | 494 | |||
FQ216483.1 | - | 3, 4 | 493 | |||
FQ216571.1 | - | 3, 4 | 494 | |||
FQ216617.1 | - | 3, 4 | 496 | |||
FQ216760.1 | - | 3, 4 | 494 | |||
FQ216828.1 | - | 3, 4 | 493 | |||
FQ217324.1 | - | 3, 4 | 494 | |||
FQ217401.1 | - | 3, 4 | 494 | |||
FQ217430.1 | - | 3, 4 | 494 | |||
FQ217483.1 | - | 3, 4 | 497 | |||
FQ217946.1 | - | 3, 4 | 495 | |||
FQ219788.1 | - | 3, 4 | 454 | |||
FQ219821.1 | - | 3, 4 | 494 | |||
FQ220371.1 | - | 3, 4 | 494 | |||
FQ220438.1 | - | 3, 4 | 454 | |||
FQ220606.1 | - | 3, 4 | 494 | |||
FQ220675.1 | - | 3, 4 | 494 | |||
FQ223760.1 | - | 3, 4 | 557 | |||
FQ224873.1 | - | 3, 4 | 496 | |||
FQ224893.1 | - | 3, 4 | 496 |
