siRNA Details
Gene Symbol: Rnf183 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006238243.2 | XP_006238305.1 | 6 | 975 | ||
XM_008763806.1 | XP_008762028.1 | 5 | 1090 | |||
XM_008763807.1 | XP_008762029.1 | 3 | 268 | |||
XM_008763808.1 | XP_008762030.1 | 5 | 753 | |||
XM_008763809.1 | XP_008762031.1 | 4 | 580 | |||
XM_008763810.1 | XP_008762032.1 | 2 | 80 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
