siRNA Details
Gene Symbol: LOC103690060 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006238497.2 | XP_006238559.1 | 4 | 1215 |
Gene Symbol: RGD1564074 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763964.1 | XP_008762186.1 | 12 | 1552 |
