siRNA Details
Gene Symbol: LOC102548397 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763971.1 | XP_008762193.1 | 1 | 1507 |
Gene Symbol: Ankrd13c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191570.1 | NP_001178499.1 | 13 | 1631 | ||
XM_006233547.2 | XP_006233609.1 | 12 | 2556 | |||
XM_008761571.1 | XP_008759793.1 | 11 | 2452 |

More Information
Other Species Targeted

Gene Ontology Categories:
