siRNA Details
Gene Symbol: Ccdc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014067.2 | NP_001014089.2 | 3 | 451 | ||
XM_006238702.2 | XP_006238764.1 | 2 | 681 | |||
XM_006238703.2 | XP_006238765.1 | 2 | 681 | |||
XM_006238704.2 | XP_006238766.1 | 3 | 609 | |||
XM_006238705.2 | XP_006238767.1 | 2 | 681 | |||
XM_006238706.2 | XP_006238768.1 | 2 | 681 | |||
XM_006238707.2 | XP_006238769.1 | 3 | 609 | |||
GenBank mRNA | BC087059.1 | - | 3 | 391 | ||
CK476945.1 | - | 3 | 397 | |||
CO385240.1 | - | 3 | 451 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
