siRNA Details
Gene Symbol: Slfnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024347.1 | NP_001019518.1 | 3 | 1288 | ||
XM_008764060.1 | XP_008762282.1 | 2 | 1597 | |||
XM_008764061.1 | XP_008762283.1 | 4 | 2822 | |||
XM_008764062.1 | XP_008762284.1 | 4 | 2822 | |||
XM_008764063.1 | XP_008762285.1 | 2 | 1597 | |||
GenBank mRNA | BC079280.1 | - | 3 | 1288 | 7134187 |
