siRNA Details
Gene Symbol: Zscan20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764175.1 | XP_008762397.1 | 3 | 757 | ||
XM_008764176.1 | XP_008762398.1 | 3 | 875 | |||
XM_008764177.1 | XP_008762399.1 | 4 | 1851 | |||
XM_008764178.1 | XP_008762400.1 | 3 | 706 | |||
XM_008764179.1 | XP_008762401.1 | 3 | 1677 | |||
XM_575911.5 | XP_575911.4 | 4 | 1434 |

More Information
Other Species Targeted

Gene Ontology Categories:
