siRNA Details
Gene Symbol: Azin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC078981.1 | - | 510 | 7112100 | ||
HQ413774.1 | - | 3 | 359 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Azin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC078981.1 | - | 510 | 7112100 | ||
HQ413774.1 | - | 3 | 359 |