siRNA Details
Gene Symbol: Zcchc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109267.1 | NP_001102737.1 | 2 | 183 | ||
XM_006238968.2 | XP_006239030.1 | 2 | 203 | |||
XM_006238969.2 | XP_006239031.1 | 2 | 197 |

More Information
Other Species Targeted

Gene Ontology Categories:
