siRNA Details
Gene Symbol: LOC500567 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024348.1 | NP_001019519.1 | 2 | 411 | ||
GenBank mRNA | BC087049.1 | - | 2 | 411 | 7112813 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
