siRNA Details
Gene Symbol: Pax7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191984.1 | NP_001178913.1 | 4 | 472 | ||
XM_006239263.2 | XP_006239325.1 | 4 | 562 |

More Information
Other Species Targeted

Gene Ontology Categories:
