siRNA Details
Gene Symbol: Szrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114599.2 | NP_001108071.1 | 4 | 388 | ||
XM_006239264.2 | XP_006239326.1 | 4 | 416 | |||
GenBank mRNA | BC083784.1 | - | 5 | 470 | ||
BP502965.1 | - | 4 | 375 | |||
FQ213371.1 | - | 5 | 489 | |||
FQ222061.1 | - | 3 | 324 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001025608, NM_001277195, BC066992, AK027934, BB871722, and others.
