siRNA Details
Gene Symbol: Fhad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750075.3 | XP_003750123.1 | 29, 30 | 3932 | ||
XM_006239329.2 | XP_006239391.1 | 29, 30 | 3932 | |||
XM_006239330.2 | XP_006239392.1 | 28, 29 | 3824 | |||
GenBank mRNA | BC098024.1 | - | 19, 20 | 2456 | 7455859 |
