siRNA Details
Gene Symbol: Tmem51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109273.1 | NP_001102743.1 | 3 | 1188 | ||
XM_006239313.2 | XP_006239375.1 | 2 | 788 | |||
XM_006239314.2 | XP_006239376.1 | 3 | 1529 | |||
XM_006239315.2 | XP_006239377.1 | 3 | 1024 | |||
XM_006239317.2 | XP_006239379.1 | 3 | 2523 | |||
XM_008764281.1 | XP_008762503.1 | 3 | 1253 |

More Information
Other Species Targeted

