siRNA Details
Gene Symbol: Morn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126086.2 | NP_001119558.1 | 4 | 351 | ||
NM_001145449.1 | NP_001138921.1 | 4 | 326 | |||
XM_006239651.2 | XP_006239713.2 | 4 | 1406 | |||
GenBank mRNA | BC166893.1 | - | 4 | 330 | 6919817 | |
CB765303.1 | - | 4 | 326 | |||
FM039224.1 | - | 4 | 314 |

More Information
Other Species Targeted

