siRNA Details
Gene Symbol: Sptssa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047743.1 | NP_001041208.1 | 1, 2 | 133 | ||
GenBank mRNA | BC098829.1 | - | 1, 2 | 133 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sptssa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047743.1 | NP_001041208.1 | 1, 2 | 133 | ||
GenBank mRNA | BC098829.1 | - | 1, 2 | 133 |