siRNA Details
Gene Symbol: Eif4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199372.2 | NP_955404.1 | 11 | 1167 | ||
XM_006246624.2 | XP_006246686.1 | 11 | 1177 | |||
GenBank mRNA | BC063812.1 | - | 11 | 1171 | 6921521 | |
FQ213709.1 | - | 5 | 409 | |||
FQ225967.1 | - | 11 | 1169 | |||
FQ233130.1 | - | 11 | 1170 | |||
FQ233795.1 | - | 3 | 232 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001159375, NM_144958, AK146464, AK162017, AK212197, and others.
Gene Ontology Categories:
