siRNA Details
Gene Symbol: Timm17al1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_576033.5 | XP_576033.1 | 2 | 180 |
Gene Symbol: Timm17a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019351.1 | NP_062224.1 | 3 | 161 | ||
GenBank mRNA | AB006450.1 | - | 3 | 161 | ||
BC061982.1 | - | 3 | 208 | 5598788 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
