siRNA Details
Gene Symbol: Cwc22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006234425.2 | XP_006234487.1 | 17 | 1946 | ||
XM_008761965.1 | XP_008760187.1 | 17 | 1961 | |||
XM_008761966.1 | XP_008760188.1 | 17 | 1948 | |||
XM_008761967.1 | XP_008760189.1 | 17 | 1958 | |||
XM_342453.7 | XP_342454.3 | 17 | 1948 | |||
GenBank mRNA | FQ231274.1 | - | 17 | 1875 | ||
FQ231579.1 | - | 17 | 1875 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
