siRNA Details
Gene Symbol: Dpf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764746.1 | XP_008762968.1 | 12 | 1488 | ||
XM_008764747.1 | XP_008762969.1 | 13 | 1469 | |||
XM_008764749.1 | XP_008762971.1 | 11 | 1317 | |||
XM_008764751.1 | XP_008762973.1 | 11 | 1734 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
