siRNA Details
Gene Symbol: Ccdc176 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006240348.2 | XP_006240410.1 | 8 | 1249 | ||
XM_008764829.1 | XP_008763051.1 | 8 | 1249 | |||
XM_008764830.1 | XP_008763052.1 | 8 | 1122 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
