siRNA Details
Gene Symbol: Efcab11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024350.1 | NP_001019521.1 | 4 | 343 | ||
XM_006240446.2 | XP_006240508.2 | 4 | 445 | |||
GenBank mRNA | BC079408.1 | - | 4 | 343 | 7113005 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
