siRNA Details
Gene Symbol: Cep170b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729727.4 | XP_002729773.2 | 12 | 3703 | ||
XM_008764990.1 | XP_008763212.1 | 12 | 4516 | |||
XM_008764991.1 | XP_008763213.1 | 13 | 4355 | |||
XM_576105.5 | XP_576105.4 | 12 | 3703 |

More Information
Other Species Targeted

