siRNA Details
Gene Symbol: Cdca4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037214.1 | NP_001032291.1 | 2 | 449 | ||
XM_008764947.1 | XP_008763169.1 | 3 | 572 | |||
XM_008764948.1 | XP_008763170.1 | 2 | 313 | |||
GenBank mRNA | BC107668.1 | - | 2 | 449 | 7937865 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
