siRNA Details
Gene Symbol: Cdca4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037214.1 | NP_001032291.1 | 2 | 429 | ||
XM_008764947.1 | XP_008763169.1 | 3 | 552 | |||
XM_008764948.1 | XP_008763170.1 | 2 | 293 | |||
GenBank mRNA | BC107668.1 | - | 2 | 429 | 7937865 |

More Information
Other Species Targeted

Gene Ontology Categories:
