siRNA Details
Gene Symbol: Ccdc105 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025774.1 | NP_001020945.1 | 7 | 1434 | ||
GenBank mRNA | BC098035.1 | - | 7 | 1434 | 7456481 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccdc105 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025774.1 | NP_001020945.1 | 7 | 1434 | ||
GenBank mRNA | BC098035.1 | - | 7 | 1434 | 7456481 |