siRNA Details
Gene Symbol: Cyp4f17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191986.1 | NP_001178915.1 | 1257 | |||
XM_006241085.2 | XP_006241147.1 | 2409 | ||||
XM_006241094.2 | XP_006241156.1 | 326 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
