siRNA Details
Gene Symbol: RGD1565498 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109287.2 | NP_001102757.2 | 3 | 1244 | ||
NM_001271273.1 | NP_001258202.1 | 3 | 1077 | |||
GenBank mRNA | FQ212465.1 | - | 3 | 1077 |
