siRNA Details
Gene Symbol: LOC500846 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047962.1 | NP_001041427.1 | 7 | 1082 | ||
GenBank mRNA | AY539897.1 | - | 7 | 1082 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
