siRNA Details
Gene Symbol: Sybu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241616.1 | XP_006241678.1 | 8 | 1782 | ||
XM_006241617.1 | XP_006241679.1 | 8 | 1779 | |||
XM_006241618.2 | XP_006241680.1 | 7 | 1942 | |||
XM_006241619.2 | XP_006241681.1 | 5 | 2709 | |||
XM_008765476.1 | XP_008763698.1 | 7 | 1723 | |||
XM_008765477.1 | XP_008763699.1 | 7 | 1723 | |||
XM_008765478.1 | XP_008763700.1 | 4 | 1098 | |||
GenBank mRNA | FQ211706.1 | - | 8 | 1779 |
