siRNA Details
Gene Symbol: CD101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256106.2 | NP_001243035.1 | 8 | 2535 | ||
NM_001256109.2 | NP_001243038.1 | 8 | 2535 | |||
NM_001256111.2 | NP_001243040.1 | 8 | 2349 | |||
NM_004258.5 | NP_004249.2 | 8 | 2535 | |||
XM_017002846.1 | XP_016858335.1 | 9 | 2488 | |||
XM_017002847.1 | XP_016858336.1 | 9 | 2488 | |||
GenBank mRNA | AK298237.1 | - | 8 | 2324 | ||
BC130327.1 | - | 9 | 2496 | 40146358 | ||
BC136484.1 | - | 9 | 2496 | 9020473 | ||
BC171768.1 | - | 9 | 2496 | 9054422 | ||
Z33642.1 | - | 9 | 2488 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
