siRNA Details
Gene Symbol: Ccdc134 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024355.1 | NP_001019526.1 | 4 | 576 | ||
XM_006242113.2 | XP_006242175.1 | 5 | 489 | |||
GenBank mRNA | BC088206.1 | - | 4 | 576 | 7381354 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
