siRNA Details
Gene Symbol: Ccdc134 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024355.1 | NP_001019526.1 | 1, 2 | 308 | ||
XM_006242113.2 | XP_006242175.1 | 2, 3 | 221 | |||
GenBank mRNA | BC088206.1 | - | 1, 2 | 308 | 7381354 |
