siRNA Details
Gene Symbol: Kdm4dl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765998.1 | XP_008764220.1 | 2 | 2276 | ||
XM_008765999.1 | XP_008764221.1 | 2 | 2279 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
