siRNA Details
Gene Symbol: RGD1561795 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109289.1 | NP_001102759.1 | 2 | 279 | ||
XM_008765967.1 | XP_008764189.1 | 4 | 618 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
