siRNA Details
Gene Symbol: Ldha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229232.2 | XP_006229294.1 | 8 | 1606 | ||
GenBank mRNA | BC060587.1 | - | 8 | 1399 | 6919582 | |
BC084698.1 | - | 7 | 1340 | 7122339 | ||
BC168737.1 | - | 8 | 1446 | 6917937 | ||
FQ222273.1 | - | 1 | 23 | |||
M54926.1 | - | 1 | 510 |
