siRNA Details
Gene Symbol: RGD1566243 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134635.1 | NP_001128107.1 | 3 | 1418 | ||
XM_006243018.2 | XP_006243080.1 | 6 | 1863 | |||
XM_006243019.2 | XP_006243081.1 | 6 | 2162 | |||
XM_008766197.1 | XP_008764419.1 | 6 | 2023 |
