siRNA Details
Gene Symbol: RGD1563941 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109293.1 | NP_001102763.1 | 1 | 139 | ||
GenBank mRNA | BC158857.1 | - | 250 | 7930388 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
