siRNA Details
Gene Symbol: RGD1562618 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113791.1 | NP_001107263.1 | 2 | 769 | ||
GenBank mRNA | BC158644.1 | - | 2 | 769 | 8363324 |

More Information
Other Species Targeted

