siRNA Details
Gene Symbol: Sh3bgrl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137647.1 | NP_001131119.1 | 2 | 292 |
Gene Symbol: LOC102554622 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006243468.2 | XP_006243530.1 | 2 | 287 |

More Information
Other Species Targeted

