siRNA Details
Gene Symbol: Mrpl41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013426.1 | NP_001013444.1 | 2 | 293 | ||
GenBank mRNA | BC091293.1 | - | 2 | 293 | 7307323 |
Gene Symbol: RGD1560917 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750552.3 | XP_003750600.1 | 1 | 329 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
