siRNA Details
Gene Symbol: RGD1560775 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766460.1 | XP_008764682.1 | 18 | 1536 | ||
XM_008766461.1 | XP_008764683.1 | 18 | 1275 | |||
XM_008766462.1 | XP_008764684.1 | 18 | 1536 | |||
XM_008766463.1 | XP_008764685.1 | 18 | 1536 | |||
XM_008766464.1 | XP_008764686.1 | 17 | 1423 | |||
XM_008766465.1 | XP_008764687.1 | 17 | 1344 | |||
XM_008766466.1 | XP_008764688.1 | 18 | 1535 | |||
GenBank mRNA | BC128758.1 | - | 17 | 1118 | 7111588 |
