siRNA Details
Gene Symbol: Bfsp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277434.1 | NP_001264363.1 | 3 | 709 | ||
GenBank mRNA | FQ228265.1 | - | 3 | 333 |

More Information
Other Species Targeted

