siRNA Details
Gene Symbol: Golga4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244126.2 | XP_006244188.1 | 4 | 584 | ||
XM_006244127.2 | XP_006244189.1 | 4 | 584 | |||
XM_006244128.2 | XP_006244190.1 | 4 | 315 | |||
XM_006244129.2 | XP_006244191.1 | 4 | 584 | |||
XM_006244130.2 | XP_006244192.1 | 4 | 584 | |||
XM_006244131.2 | XP_006244193.1 | 3 | 485 | |||
XM_006244132.2 | XP_006244194.1 | 4 | 584 | |||
XM_006244133.2 | XP_006244195.1 | 3 | 485 |
