siRNA Details
Gene Symbol: Golga4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191080.1 | NP_001178009.1 | 14 | 2800 | ||
XM_006244126.2 | XP_006244188.1 | 16 | 3320 | |||
XM_006244127.2 | XP_006244189.1 | 16 | 3290 | |||
XM_006244128.2 | XP_006244190.1 | 16 | 3051 | |||
XM_006244129.2 | XP_006244191.1 | 16 | 3320 | |||
XM_006244130.2 | XP_006244192.1 | 15 | 3236 | |||
XM_006244131.2 | XP_006244193.1 | 15 | 3221 | |||
XM_006244132.2 | XP_006244194.1 | 16 | 3320 | |||
XM_006244133.2 | XP_006244195.1 | 15 | 3221 |
