siRNA Details
Gene Symbol: Pdcd6ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029910.3 | NP_001025081.2 | 13 | 1798 | ||
XM_006243997.1 | XP_006244059.1 | 13 | 2133 | |||
GenBank mRNA | AF192757.1 | - | 3 | 265 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
