siRNA Details
Gene Symbol: Sult1c2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013177.2 | NP_001013195.2 | 857 | |||
GenBank mRNA | AJ238392.1 | - | 798 | |||
BC081691.1 | - | 754 | 7110136 | |||
BC092564.1 | - | 857 | 7372924 | |||
FQ209913.1 | - | 851 |
Gene Symbol: RGD1559960 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244256.2 | XP_006244318.2 | 4 | 541 | ||
XM_008766792.1 | XP_008765014.1 | 5 | 688 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
