siRNA Details
Gene Symbol: Lrrc73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024360.2 | NP_001019531.1 | 5 | 773 | ||
XM_008766878.1 | XP_008765100.1 | 4 | 1749 | |||
GenBank mRNA | AB210855.1 | - | 5 | 673 | ||
BC128712.1 | - | 5 | 773 | 7133504 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001111142, BC147714, AK134769.
