siRNA Details
Gene Symbol: Bnip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053420.3 | NP_445872.1 | 4 | 441 | ||
GenBank mRNA | AF243515.1 | - | 4 | 405 | ||
AY095482.1 | - | 4 | 405 | |||
BC070958.1 | - | 4 | 399 | 7105707 | ||
BC081690.1 | - | 4 | 379 | 7190121 | ||
CO403082.1 | - | 4 | 477 | |||
FQ213448.1 | - | 3 | 202 | |||
FQ215108.1 | - | 4 | 411 | |||
FQ215127.1 | - | 4 | 411 | |||
FQ215203.1 | - | 4 | 432 | |||
FQ216745.1 | - | 4 | 432 | |||
FQ216768.1 | - | 1 | 18 | |||
FQ216807.1 | - | 4 | 432 | |||
FQ220853.1 | - | 4 | 410 | |||
FQ221849.1 | - | 4 | 432 | |||
KC660101.1 | - | 3 | 238 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009760, AK170865, AK075943, AF041054, AK014223, and others.
Gene Ontology Categories:
