siRNA Details
Gene Symbol: Als2cr11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024363.1 | NP_001019534.2 | 12, 13 | 1327 | ||
GenBank mRNA | BC083917.1 | - | 12, 13 | 1327 | 7134752 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
